ENA332672033 Enamine

methyl 4-bromo-5-formyl-1-methyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD19381978
  • Cas Number : 1315365-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79382

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79382
Availability details for
Quantity
  • Availability
  • Add to Cart