ENA332672034 Enamine

1-(pyridazin-3-yl)piperidin-3-amine dihydrochloride

  • MDL number MFCD19381979
  • Cas Number : 1315366-14-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79383

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79383
Availability details for
Quantity
  • Availability
  • Add to Cart