ENA332672058 Enamine

pent-4-en-2-amine hydrochloride

  • MDL number MFCD19382008
  • Cas Number : 1315366-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77...79
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79544

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-79544
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart