ENA332672087 Enamine

1-{[5-(propan-2-yl)-1,2,4-oxadiazol-3-yl]methyl}-1,4-diazepane dihydrochloride

  • MDL number MFCD19382044
  • Cas Number : 1315368-78-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79681

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79681
Availability details for
Quantity
  • Availability
  • Add to Cart