ENA332672088 Enamine

1-[(5-cyclopropyl-1,2,4-oxadiazol-3-yl)methyl]-1,4-diazepane dihydrochloride

  • MDL number MFCD19382045
  • Cas Number : 1315368-72-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176...178
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79683

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79683
Availability details for
Quantity
  • Availability
  • Add to Cart