ENA332672102 Enamine

(1S)-1-(1,3-thiazol-5-yl)ethan-1-ol

  • MDL number MFCD19382062
  • Cas Number : 1315378-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79769

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79769
Availability details for
Quantity
  • Availability
  • Add to Cart