ENA332672109 Enamine

(oxan-4-ylmethyl)hydrazine dihydrochloride

  • MDL number MFCD19382069
  • Cas Number : 1315365-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145...147
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79797

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79797
Availability details for
Quantity
  • Availability
  • Add to Cart