ENA332672122 Enamine

6-[(piperazin-1-yl)carbonyl]-1,3-benzoxazol-2-amine

  • MDL number MFCD19382082
  • Cas Number : 1315368-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79835

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79835
Availability details for
Quantity
  • Availability
  • Add to Cart