ENA332672162 Enamine

1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-3-methyl-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD19382124
  • Cas Number : 1315368-04-7
  • Molecular Weight : 373.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-3-methyl-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
Inchi Code 1S/C12H6ClF6N3O2/c1-4-7(10(23)24)8(12(17,18)19)22(21-4)9-6(13)2-5(3-20-9)11(14,15)16/h2-3H,1H3,(H,23,24)
Inchi Key WZIXWCFGCYJKKD-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-79968

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79968
Availability details for
Quantity
  • Availability
  • Add to Cart