ENA332672167 Enamine

2-(diethylamino)butanoic acid hydrochloride

  • MDL number MFCD19382129
  • Cas Number : 1315365-38-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-79984

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79984
Availability details for
Quantity
  • Availability
  • Add to Cart