ENA332672175 Enamine

2-(chloromethyl)-4-(propan-2-yl)morpholine

  • MDL number MFCD19382137
  • Cas Number : 48119-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80002

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80002
Availability details for
Quantity
  • Availability
  • Add to Cart