ENA332672194 Enamine

2-(6-oxopiperidin-2-yl)acetamide

  • MDL number MFCD19382158
  • Cas Number : 1071661-20-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80076

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80076
Availability details for
Quantity
  • Availability
  • Add to Cart