ENA332672226 Enamine

benzyl 3-(methanesulfonyloxy)azetidine-1-carboxylate

  • MDL number MFCD19382192
  • Cas Number : 1315368-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80213

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80213
Availability details for
Quantity
  • Availability
  • Add to Cart