ENA332672232 Enamine

2-(1H-1,2,4-triazol-1-ylmethyl)morpholine hydrochloride

  • MDL number MFCD19382199
  • Cas Number : 1315366-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 239...241
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80241

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80241
Availability details for
Quantity
  • Availability
  • Add to Cart