ENA332672234 Enamine

[1-(pyridazin-3-yl)pyrrolidin-2-yl]methanamine dihydrobromide

  • MDL number MFCD19382201
  • Cas Number : 1315366-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 222...224
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80260

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80260
Availability details for
Quantity
  • Availability
  • Add to Cart