ENA332672236 Enamine

ethyl 1-ethyl-5-oxo-4,5-dihydro-1H-pyrazole-3-carboxylate

  • MDL number MFCD19382203
  • Cas Number : 1315368-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135...137
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80266

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80266
Availability details for
Quantity
  • Availability
  • Add to Cart