ENA332672239 Enamine

3-(3-aminopropyl)-1-phenyl-1H-pyrazol-5-amine dihydrochloride

  • MDL number MFCD19382207
  • Cas Number : 1315367-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Product Link https://www.enaminestore.com/catalog/EN300-80288

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80288
Availability details for
Quantity
  • Availability
  • Add to Cart