ENA332672246 Enamine

6-methyl-N-(piperidin-4-yl)pyridazin-3-amine dihydrobromide

  • MDL number MFCD19382214
  • Cas Number : 1315365-29-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80340

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80340
Availability details for
Quantity
  • Availability
  • Add to Cart