ENA332672248 Enamine

5-(chlorodifluoromethyl)-1-(3-chloropyridin-2-yl)-3-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD19382217
  • Cas Number : 1315366-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80350

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80350
Availability details for
Quantity
  • Availability
  • Add to Cart