ENA332672249 Enamine

4-(cyclopropylmethoxy)-2-fluoroaniline hydrochloride

  • MDL number MFCD19382218
  • Cas Number : 1315369-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80383

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80383
Availability details for
Quantity
  • Availability
  • Add to Cart