ENA332672254 Enamine

1',2'-dihydrospiro[cyclopentane-1,3'-indole]

  • MDL number MFCD12030572

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79344

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79344
Availability details for
Quantity
  • Availability
  • Add to Cart