ENA332672258 Enamine

quinoxaline-6-carbonitrile

  • MDL number MFCD08669502
  • Cas Number : 23088-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77906

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-77906
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart