ENA332672261 Enamine

3,4,5,6-tetrahydro-2H-azepin-7-amine hydrochloride

  • MDL number MFCD00466070
  • Cas Number : 54661-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77981

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77981
Availability details for
Quantity
  • Availability
  • Add to Cart