ENA332672268 Enamine

2-chloro-N-cyclopropyl-N-[(4-methylphenyl)methyl]acetamide

  • MDL number MFCD11919335
  • Cas Number : 1181747-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-47359

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47359
Availability details for
Quantity
  • Availability
  • Add to Cart