ENA332672287 Enamine

4-bromopyridin-1-ium-1-olate

  • MDL number MFCD00234039
  • Cas Number : 14248-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-144
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79462

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79462
Availability details for
Quantity
  • Availability
  • Add to Cart