ENA332672292 Enamine

2-acetamido-1,3-thiazole-5-sulfonyl chloride

  • MDL number MFCD05022486
  • Cas Number : 69812-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79472

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-79472
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart