ENA332672297 Enamine

2-methyl-1,3-benzothiazole-6-sulfonyl chloride

  • MDL number MFCD03990571
  • Cas Number : 21431-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98-101
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79478

Safety Information

signalword Danger
MSDS https://www.enaminestore.com/catalog/EN300-79478
pictograms GHS05
hstatements EUH014,H314
Availability details for
Quantity
  • Availability
  • Add to Cart