ENA332672315 Enamine

4-chloro-2-methyl-5-nitroaniline

  • MDL number MFCD00007843
  • Cas Number : 34648-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78806

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78806
Availability details for
Quantity
  • Availability
  • Add to Cart