ENA332672331 Enamine

3-(4-methoxybenzenesulfonyl)-2-methylpropanoic acid

  • MDL number MFCD10024399
  • Cas Number : 205504-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-79532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79532
Availability details for
Quantity
  • Availability
  • Add to Cart