ENA332672339 Enamine

3,3-dimethyl-5-(morpholin-4-yl)-5-oxopentanoic acid

  • MDL number MFCD11851195
  • Cas Number : 879131-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79542

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79542
Availability details for
Quantity
  • Availability
  • Add to Cart