ENA332672342 Enamine

N-[2-(4-fluorophenyl)ethyl]cyclopropanamine

  • MDL number MFCD11212566
  • Cas Number : 625435-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79545

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79545
Availability details for
Quantity
  • Availability
  • Add to Cart