ENA332672352 Enamine

3-cyclopropyl-6-hydrazinylpyridazine

  • MDL number MFCD16871610
  • Cas Number : 1315366-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79556

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79556
Availability details for
Quantity
  • Availability
  • Add to Cart