ENA332672370 Enamine

2-(2,5-dioxoimidazolidin-4-yl)ethane-1-sulfonyl chloride

  • MDL number MFCD19200124
  • Cas Number : 98021-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79583

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79583
Availability details for
Quantity
  • Availability
  • Add to Cart