ENA332672380 Enamine

2,6-difluoro-4-methoxybenzene-1-sulfonyl chloride

  • MDL number MFCD12813878
  • Cas Number : 1178498-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60114

Safety Information

signalword Danger
pcodes P260,P264,P270,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P330,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-60114
pictograms GHS05,GHS07
hstatements H302,H314
Availability details for
Quantity
  • Availability
  • Add to Cart