ENA332672382 Enamine

8-chloro-3-methyl-[1,2,4]triazolo[4,3-a]pyrazine

  • MDL number MFCD08272087
  • Cas Number : 68774-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 212...214
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62151

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62151
Availability details for
Quantity
  • Availability
  • Add to Cart