ENA332672399 Enamine

ethyl 4-(trifluoromethyl)benzene-1-carboximidate

  • MDL number MFCD09751973
  • Cas Number : 57870-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79616

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79616
Availability details for
Quantity
  • Availability
  • Add to Cart