ENA332672402 Enamine

3-(cyclopropylamino)propan-1-ol

  • MDL number MFCD15206831
  • Cas Number : 1211444-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79620

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-79620
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart