ENA332672405 Enamine

(1-bromopropan-2-yl)benzene

  • MDL number MFCD00017888
  • Cas Number : 1459-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79625

Safety Information

signalword Danger
pcodes P261,P264,P270,P285,P301+P312,P304+P341,P330,P342+P311,P501
hcodes Xn
MSDS https://www.enaminestore.com/catalog/EN300-79625
pictograms GHS07,GHS08
hstatements H302,H334
Availability details for
Quantity
  • Availability
  • Add to Cart