ENA332672412 Enamine

4-(2-oxo-1,2-dihydropyridin-1-yl)butanoic acid

  • MDL number MFCD09811490
  • Cas Number : 71644-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79634

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79634
Availability details for
Quantity
  • Availability
  • Add to Cart