ENA332672414 Enamine

2,5-dibromothiophene-3-carboxylic acid

  • MDL number MFCD08445656
  • Cas Number : 7311-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177-179
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79638

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79638
Availability details for
Quantity
  • Availability
  • Add to Cart