ENA332672415 Enamine

6-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-3-amine

  • MDL number MFCD18261251
  • Cas Number : 1211578-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79639

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-79639
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart