ENA332672454 Enamine

3-(4-fluorophenyl)-3-methylbutan-2-one

  • MDL number MFCD09904319
  • Cas Number : 1017409-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-79700

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79700
Availability details for
Quantity
  • Availability
  • Add to Cart