ENA332672455 Enamine

1-(2-methoxyethyl)-2-methylpiperazine

  • MDL number MFCD16300980
  • Cas Number : 1225603-87-1
  • Molecular Weight : 158.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-methoxyethyl)-2-methylpiperazine
Inchi Code 1S/C8H18N2O/c1-8-7-9-3-4-10(8)5-6-11-2/h8-9H,3-7H2,1-2H3
Inchi Key WGOQNROQRIISTQ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70952

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70952
Availability details for
Quantity
  • Availability
  • Add to Cart