ENA332672475 Enamine

[2-(4-chlorophenyl)ethyl](cyclopropylmethyl)amine

  • MDL number MFCD12109151
  • Cas Number : 1156283-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-79733

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79733
Availability details for
Quantity
  • Availability
  • Add to Cart