ENA332672479 Enamine

7-chloro-[1,2,4]triazolo[1,5-a]pyridin-2-amine

  • MDL number MFCD11870488
  • Cas Number : 1131410-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79738

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79738
Availability details for
Quantity
  • Availability
  • Add to Cart