ENA332672490 Enamine

3-[(2-methoxyethanesulfinyl)methyl]aniline

  • MDL number MFCD14631029
  • Cas Number : 1250234-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-79749

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79749
Availability details for
Quantity
  • Availability
  • Add to Cart