ENA332672500 Enamine

1-(1,3-thiazol-5-yl)ethan-1-amine

  • MDL number MFCD10007038
  • Cas Number : 1017480-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79766

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79766
Availability details for
Quantity
  • Availability
  • Add to Cart