ENA332672503 Enamine

1-phenoxy-3-(piperazin-1-yl)propan-2-ol

  • MDL number MFCD00218466
  • Cas Number : 40944-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71174

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71174
Availability details for
Quantity
  • Availability
  • Add to Cart