ENA332672520 Enamine

2-chloro-N-[1-(4-ethylphenyl)ethyl]propanamide

  • MDL number MFCD12147727
  • Cas Number : 1153022-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-28055

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28055
Availability details for
Quantity
  • Availability
  • Add to Cart