ENA332672531 Enamine

3-phenyl-1H-1,2,4-triazole

  • MDL number MFCD00159717
  • Cas Number : 3357-42-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117-121
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-79821

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79821
Availability details for
Quantity
  • Availability
  • Add to Cart